Praseodymium ≥99.9% (REO, rare earth oxide basis), powder -200 mesh
Supplier: Thermo Scientific Chemicals
|
Danger
|
45910-06
45910-14
CAAA45910-06EA
395
CAD
CAAA45910-06
CAAA45910-14
Praseodymium ≥99.9% (REO, rare earth oxide basis), powder -200 mesh
Praseodymium
- Packaged under argon
Purity: 99.9%
CAS No.: 7440-10-0
Physical Form: Packaged under argon
Sensitivity: Air and Moisture Sensitive
Appearance: Silver
Odor: Odorless
UN No.: 3089
MDL No.: MFCD00011174
Formula:
Pr Storage Temperature: Ambient |
MDL Number:
MFCD00011174 CAS Number: 7440-10-0 EINECS: 231-120-3 UN: 3089 ADR: 4.1,II |
Specification Test Results
Assay Minimum | 99.9 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...