Magnesium Oxide Crucibles, Cylindrical with Flat Base, Thermo Scientific
Supplier: Thermo Scientific Chemicals
42991-KF 45680-KS 45733-KS 45854-KS 45900-KS
CAAA42989-KFEA 501.22 CAD
CAAA42989-KF CAAA42989-KJ CAAA42991-KF CAAA42991-KJ CAAA45680-KS CAAA45733-KS CAAA45854-KS CAAA45900-KS
Magnesium Oxide Crucibles, Cylindrical with Flat Base, Thermo Scientific
Crucibles
These high performance oxide ceramics display superior characteristics both at elevated temperature and at high frequencies, and are resistant to corrosive liquids and gases.
  • Cylindrical, flat bottom design
  • Crucibles provide increased working temperature characteristics
  • Manufactured of pure metal oxides without the admixtures of silicates
Oxide ceramics are also inert to oxidation and are not subject to radiation damage.