Trays, Rectangular, Thermo Scientific
Supplier: Thermo Scientific
045634-KS 045791-KS 45699-KS 45954-KS 045865-KS 045827-KS 045749-KS 45699-KS 45749-KS 45634-KS 45827-KS 45791-KS 45865-KS 45954-KS
CAAA45865-KSEA 1033.32 CAD
CAAA45865-KS AA45791-KS AA45749-KS AA45634-KS AA45954-KS AA45827-KS AA45699-KS AA45865-KS CAAA45827-KS CAAA45634-KS CAAA45791-KS CAAA45749-KS CAAA45954-KS CAAA45699-KS
Trays, Rectangular, Thermo Scientific
Trays/Dishes/Pots Laboratory Trays/Dishes
These rectangular trays are made up of high purity magnesia (MgO).
  • Stability at high temperature and high frequencies
  • Resistant to corossive liquids and gases
  • Ultra high vacuum resistance
  • Extreme purity
  • Thermal shock resistance
  • Inert to oxidation and radiation damage