Lead in Isooctane Standard Solution, Specpure®, Thermo Scientific

Supplier: Thermo Scientific Chemicals

45325-22 45326-22 45327-22 45328-22 45329-22 45330-22 45332-22 45333-22 45334-22 45335-22 045336-22 045354-22 045331-22
CAAA45325-22EA 216.46 CAD
CAAA45325-22 CAAA45326-22 CAAA45327-22 CAAA45328-22 CAAA45329-22 CAAA45330-22 CAAA45332-22 CAAA45333-22 CAAA45334-22 CAAA45335-22 AA45336-22 AA45354-22 AA45331-22
Lead in Isooctane Standard Solution, Specpure®, Thermo Scientific
Standards Petroleum Industry Standards

Molecular Formula: Matrix: Isooctane
Physical Form: Liquid
Odor: Nearly odorless
UN No.: 1262
MDL No.: MFCD01094200

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR