Germanium, plasma standard solution, Specpure®, Ge 10µg/ml, Thermo Scientific

Supplier: Thermo Scientific
Specpure®
45254-AE 045254-AE
CAAA45254-AEEA 115.34 CAD
AA45254-AE CAAA45254-AE
Germanium, plasma standard solution, Specpure®, Ge 10µg/ml, Thermo Scientific
Standards Cationic Standards Germanium Standards
Liquid

Molecular Formula: Ge in 2% HNO3/tr. HF
Physical Form: Liquid
UN No.: 3264
MDL No.: MFCD00085310
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR