Inconel® 600 alloy, gauze woven from wire 100 mesh Ø 0.102 mm (0.004 in)
Supplier: Thermo Scientific Chemicals
|
Danger
|
45069-GH
45069-GT
CAAA45069-FIEA
73.36
CAD
CAAA45069-FI
CAAA45069-GH
CAAA45069-GT
Inconel® 600 alloy, gauze woven from wire 100 mesh Ø 0.102 mm (0.004 in)
Inconel® 600 alloy
Molecular Formula: Ni:Cr:Fe; 77:15.5:7.5 wt%
MDL No.: MFCD01741927
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...