Hydrochloric acid standardised solution 6 mol/l (6 N)

Supplier: Thermo Scientific Chemicals
Danger

Synonyms: Hydrochloric acid solution, Muriatic acid

44921-K2 44921-K7 44921-M4
CAAA44921-K2EA 83.58 CAD
CAAA44921-K2 CAAA44921-K7 CAAA44921-M4
Hydrochloric acid standardised solution 6 mol/l (6 N)
Hydrochloric acid

  • Liquid


CAS No.: 7647-01-0 Molecular Formula: HCl Formula Weight: 36.46 Physical Form: Liquid Appearance: Colorless to pale yellow UN No.: 1789 MDL No.: MFCD00011324

Formula: HCl
MW: 36.46
Storage Temperature: Ambient
MDL Number: MFCD00011324
CAS Number: 7647-01-0
EINECS: 231-595-7
UN: 1789
ADR: 8 II

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR