HSG-75, Electrically conductive heat sink grease, Ag ≈75%, Thermo Scientific

Supplier: Thermo Scientific
44590-MJ 44590-MJ 44590-O0 044590-O0
CAAA44590-O0EA 276.5 CAD
CAAA44590-O0 AA44590-O0 CAAA44590-MJ AA44590-MJ
HSG-75, Electrically conductive heat sink grease, Ag ≈75%, Thermo Scientific
Lubricants
Thermally and electrically conductive heat sink grease applied to the base and mounting stud of transistors, diodes and rectifiers, as a coupling agent to reduce thermal and electrical contact resistance

Physical Form: Viscous liquid
Appearance: Grey
UN No.:UN3082

Applications: A high loading of proprietary silver flake provides excellent electrical conductivity properties. This is ideal for mounting sputter targets where thermal or thermal plus electrical conductivity are needed.

Ideal for sputtering target applications
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR