Tantalum, gauze woven from wire 30 mesh woven from 0.076mm (0.003in)
Supplier: Thermo Scientific Chemicals
043965-GN
043965-GY
43965-GN
43965-GY
AA43965-GNEA
548.26
CAD
AA43965-GN
AA43965-GY
CAAA43965-GN
CAAA43965-GY
Tantalum, gauze woven from wire 30 mesh woven from 0.076mm (0.003in)
Tantalum
- Wire Cloth
CAS No.: 7440-25-7
Physical Form: Wire Cloth
MDL No.: MFCD00011252
Formula:
Ta Storage Temperature: Ambient |
MDL Number:
MFCD00011252 CAS Number: 7440-25-7 EINECS: 231-135-5 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...