Zinc ≥99.999% (metals basis), foil, Puratronic®, Thickness: 2 mm (0.08 in)

Supplier: Thermo Scientific Chemicals

43504-FF 043504-FI 43504-FI
CAAA43504-FFEA 769.46 CAD
CAAA43504-FF AA43504-FI CAAA43504-FI
Zinc ≥99.999% (metals basis), foil, Puratronic®, Thickness: 2 mm (0.08 in)
Zinc

  • ~8.9g/25x25mm


Purity: 99.999% CAS No.: 7440-66-6 Physical Form: ≈8.9g/25x25mm Odor: Odorless MDL No.: MFCD00011291

Formula: Zn
Storage Temperature: Ambient
MDL Number: MFCD00011291
CAS Number: 7440-66-6
EINECS: 231-175-3

Order Now

Specification Test Results

Assay Minimum 99.999


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR