Carbon ≥99%, felt, Thickness: 6.35 mm (0.250 in)
Supplier: Thermo Scientific Chemicals
Synonyms:
Charcoal
043200-RF
043200-RR
43200-RF
43200-RR
AA43200-RFEA
148.25
CAD
AA43200-RF
AA43200-RR
CAAA43200-RF
CAAA43200-RR
Carbon ≥99%, felt, Thickness: 6.35 mm (0.250 in)
Carbon
~4.8g/10x10cm
Purity: 99.0%
Physical Form: ≈4.8g/10x10cm
MDL No.: MFCD00133992
Formula:
C MW: 12.01 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD00133992 CAS Number: 7440-44-0 EINECS: 231-955-3 |
Specification Test Results
Assay Minimum | 99 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...