Beakers, Copper, Thermo Scientific
Supplier: Thermo Scientific Chemicals
039052-KT 039053-KT 039056-KT
AA39052-KTEA 214.88 CAD
AA39052-KT AA39053-KT AA39054-KT AA39056-KT
Beakers, Copper, Thermo Scientific
Beakers
These beakers are made up of copper.
  • Provides high electrical and thermal conductivities
  • High corrosive resistance
  • Low toxicity
Copper is considered as one of the most important metallic elements, due to its unique physical and chemical properties. With similar reactivity to silver and gold, it is classified as a noble metal.