Combustion Boats, Glassy Carbon, Thermo Scientific
Supplier: Thermo Scientific Chemicals
39028-KT 039026-KT 039027-KT 039028-KT
CAAAA39028-KTEA 695.2 CAD
CAAAA39028-KT AA39026-KT AA39027-KT AA39028-KT
Combustion Boats, Glassy Carbon, Thermo Scientific
Boats Combustion Boats
Combustion boats, glassy carbon.
  • Resistance to all wet decomposition agents
  • No memory effects (uncontrolled adsorption and release of foreign elements)
  • No contamination of analytical samples
  • Stability to acid and alkaline melts
  • No wetting effect of metal melts
  • Resistive to abrasive wear
  • Good electrical conductivity
Boats for high-temperature differential thermal/thermal gravimetric analyses. Good resistance to thermal shock allows rapid heating and cooling times.