Glassy Carbon Cylindrical Crucibles, Thermo Scientific
Supplier: Thermo Scientific Chemicals
039008-KT 039010-KT 039011-KT 039012-KT 039014-KT
AA39008-KTEA 729.96 CAD
AA39008-KT AA39010-KT AA39011-KT AA39012-KT AA39014-KT
Glassy Carbon Cylindrical Crucibles, Thermo Scientific
Crucibles
Glassy carbon offers high purity, corrosion resistance, thermal stability, and a structure impermeable to both gases and liquids
  • Resistance to all wet decomposition agents
  • No memory effects (uncontrolled adsorption and release of foreign elements)
  • No contamination of analytical samples
  • Stability to acid and alkaline melts
  • No wetting effect of metal melts
  • Good resistance to thermal shock allows rapid heating and cooling times
  • Resistive to abrasive wear
  • Good electrical conductivity
These crucibles are designed for high-temperature differential thermal/More Product Information