Dishes, Evaporating, Glassy Carbon, Thermo Scientific
Supplier: Thermo Scientific
38372-KT
AA38372-KTEA 1433.06 CAD
AA38372-KT
Dishes, Evaporating, Glassy Carbon, Thermo Scientific
Trays/Dishes/Pots Evaporating Basins/Dishes
These evaporating dishes are made up of glassy carbon.
  • Resistance to all wet decomposition agents
  • No memory effects (uncontrolled adsorption and release of foreign elements)
  • No contamination of analytical samples
  • Stability to acid and alkaline melts
  • No wetting effect of metal melts
  • Good resistance to thermal shock allows rapid heating and cooling times
  • Resistive to abrasive wear
  • Good electrical conductivity
⁴/₄ ⁴/₄ -1 -1