Bottles, Wide Neck, PTFE, with Screw Cap, Thermo Scientific
Supplier: Thermo Scientific Chemicals
038111-KT 038112-KT 038113-KT 038114-KT 038115-KT
AA38110-KTEA 237.43 CAD
AA38110-KT AA38111-KT AA38112-KT AA38113-KT AA38114-KT AA38115-KT
Bottles, Wide Neck, PTFE, with Screw Cap, Thermo Scientific
Bottles Plastic Bottles
PTFE, natural, with screw cap.
  • Inert
  • Heat resistant up to +280 °C
Ideal for storage and transport of valuable or aggressive materials. It features heavy wall construction with super smooth internal finish.