Watch Glasses/Beaker Cover, PTFE, Thermo Scientific
Supplier: Thermo Scientific Chemicals
038093-KT 038095-KT 038096-KT 038097-KT 038098-KT 038099-KT
AA38092-KTEA 29.86 CAD
AA38092-KT AA38093-KT AA38094-KT AA38095-KT AA38096-KT AA38097-KT AA38098-KT AA38099-KT
Watch Glasses/Beaker Cover, PTFE, Thermo Scientific
Watch Glasses
These covers are manufactured from pure PTFE
Used to cover beakers during digestions and for spotting out.