Molecular sieve 13X (1.0 nm, 10 Å), pellets, Ø 3-4mm (0.12-0.16in)
Supplier: Thermo Scientific Chemicals
033550-A1
033550-A7
AA33550-A1EA
142.39
CAD
AA33550-A1
AA33550-A7
Molecular sieve 13X (1.0 nm, 10 Å), pellets, Ø 3-4mm (0.12-0.16in)
Molecular sieve 13X (1.0 nm, 10 Å)
1/8in Pellets
CAS No.: 63231-69-6
Physical Form: 1/8in Pellets
Sensitivity: Hygroscopic
MDL No.: MFCD00131613
Storage Temperature:
Ambient |
MDL Number:
MFCD00164506 CAS Number: 63231-69-6 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...