Molecular sieve 3A (0.3 nm, 3 Å), pellets, Ø 3-4mm (0.12-0.16in)
Supplier: Thermo Scientific Chemicals
Synonyms:
Molecular sieve 0.3 nm, 3A molecular sieve, Molecular sieve A3, Linde 3A, Sodium aluminium silicate
033530-A1
033530-A7
AA33530-A1EA
235.42
CAD
AA33530-A1
AA33530-A7
Molecular sieve 3A (0.3 nm, 3 Å), pellets, Ø 3-4mm (0.12-0.16in)
Molecular sieve 3A (0.3 nm, 3 Å)
Pellets
CAS No.: 308080-99-1
Physical Form: Pellets
Sensitivity: Hygroscopic
Appearance: Pale brown
MDL No.: MFCD00147627
Formula:
KnNa₁₂-n[(AlO₂)₁₂(SiO₂)₁₂]·xH₂O Storage Temperature: Ambient |
MDL Number:
MFCD00131613 CAS Number: 308080-99-1 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...