Molecular sieve A4 (0.4 nm, 4 Å), beads -8+12 (∼ 2mm), with indicator
Supplier: Thermo Scientific Chemicals
|
Danger
|
Synonyms:
Molecular sieve 0.4 nm, Molecular sieves pack 4A, Linde 4A, Sodium aluminium silicate
032256-30
032256-A1
032256-A7
AA32256-30EA
132.86
CAD
AA32256-30
AA32256-A1
AA32256-A7
Molecular sieve A4 (0.4 nm, 4 Å), beads -8+12 (∼ 2mm), with indicator
Molecular sieve A4 (0.4 nm, 4 Å)
-8+12 Beads
CAS No.: 70955-01-0
Physical Form: -8+12 Beads
Sensitivity: Hygroscopic
Appearance: Blue
Odor: Odorless
UN No.: 3077
MDL No.: MFCD00147627
Storage Temperature:
Ambient |
MDL Number:
MFCD00147627 CAS Number: 70955-01-0 UN: 3077 ADR: 9,III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...