Lutetium trinitrate hydrate ≥99.9% (REO, rare earth oxide basis)

Supplier: Thermo Scientific Chemicals
Danger

14568-14 14568-03 14568-06
CAAAA14568-14EA 1169.2 CAD
CAAAA14568-14 CAAA14568-03 CAAA14568-06
Lutetium trinitrate hydrate ≥99.9% (REO, rare earth oxide basis)
Lutetium trinitrate hydrate

  • Crystalline Aggregates


Purity: 99.9% CAS No.: 100641-16-5 Molecular Formula: LuN3O9.xH2O Formula Weight: 360.98(anhy) Physical Form: Crystalline Aggregates Sensitivity: Hygroscopic Appearance: White UN No.: 1477 MDL No.: MFCD00150421

Formula: LuN₃O₉·H₂O
Storage Temperature: Ambient
MDL Number: MFCD00150421
CAS Number: 100641-16-5
EINECS: 233-241-7
UN: 1477
ADR: 5.1,III

Order Now

Specification Test Results

Assay Minimum 99.9


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR