Rubidium chloride ≥99%
Supplier: Thermo Scientific Chemicals
Warning
|
Synonyms:
Rubidiumchloride
Testing for perchloric acid, source of rubidium, catalyst
- Crystalline
Purity: 99%
CAS No.: 7791-11-9
Molecular Formula: RbCl
Formula Weight: 120.92
Physical Form: Crystalline
Density: 2.800
Refractive Index: 1.493
Sensitivity: Hygroscopic
Appearance: White
Odor: Odorless
Melting Point: 718°
Boiling Point: 1390°
Solubility: Soluble in water. Practically insoluble in alcohol
MDL No.: MFCD00011187
Applications: Testing for perchloric acid, source of rubidium, catalyst
Formula:
RbCl Boiling Pt: 1390 °C Melting Pt: 718 °C Density: 2.800 Storage Temperature: Ambient |
MDL Number:
MFCD00011187 CAS Number: 7791-11-9 EINECS: 232-240-9 Merck Index: 14,8284 |
Specification Test Results
Assay Minimum | 99 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...