Lithium sulfide ≥99.9% (metals basis)
Supplier: Thermo Scientific Chemicals
|
Danger
|
12839-04
12839-09
12839-18
012839-30
CAAA12839-04EA
88.8
CAD
CAAA12839-04
CAAA12839-09
CAAA12839-18
AA12839-30
Lithium sulfide ≥99.9% (metals basis)
Lithium sulphide
- -200 Mesh Powder
Purity: 99.9%
CAS No.: 12136-58-2
Molecular Formula: Li2S
Formula Weight: 45.95
Physical Form: -200 Mesh Powder
Density: 1.66
Sensitivity: Moisture Sensitive
Appearance: Yellow
Odor: Like rotten eggs
Melting Point: 900-975°
UN No.: 3134
MDL No.: MFCD00011085
Formula:
Li₂S Melting Pt: 900…975 °C Density: 1.66 Storage Temperature: Ambient |
MDL Number:
MFCD00011085 CAS Number: 12136-58-2 EINECS: 235-228-1 UN: 3134 ADR: 4.3,II |
Specification Test Results
Assay Minimum | 99.9 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...