Niobium ≥99.8% (metals basis), pieces
Supplier: Thermo Scientific Chemicals
00241-22
000241-36
00241-36
CAAA00241-22EA
172.22
CAD
CAAA00241-22
AA00241-36
CAAA00241-36
Niobium ≥99.8% (metals basis), pieces
Niobium
Purity: 99.8%
CAS No.: 7440-03-1
Appearance: Grey or silver
Odor: Odorless
MDL No.: MFCD00011126
Formula:
Nb MW: 92.91 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD00011126 CAS Number: 7440-03-1 EINECS: 231-113-5 |
Specification Test Results
Assay Minimum | 99.8 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...