2-Fluoro-6-iodobenzoic acid 97%
Supplier: Thermo Scientific Chemicals
|
Warning
|
L07924-03
L07924-06
CAAAL07924-03EA
70.78
CAD
CAAAL07924-03
CAAAL07924-06
2-Fluoro-6-iodobenzoic acid 97%
2-Fluoro-6-iodobenzoic acid
CAS: 111771-08-5
MDL: MFCD00042289
Formula:
C₇H₄FIO₂ Boiling Pt: 312 °C (1013 hPa) Melting Pt: 122…126 °C Storage Temperature: Ambient |
MDL Number:
MFCD00042289 CAS Number: 111771-08-5 |
Specification Test Results
Assay Minimum | 97 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...