Ethyl chloroformate 97%
Supplier: Thermo Scientific Chemicals
|
Danger
|
Synonyms:
Chloroformic acid ethyl ester
L06311-06
L06311-06
L06311-22
L06311-36
CAAAAL06311-06EA
39.03
CAD
CAAAAL06311-06
AAL06311-06
AAL06311-22
AAL06311-36
Ethyl chloroformate 97%
Ethyl chloroformate
CAS: 541-41-3
MDL: MFCD00000644
Formula:
C₃H₅ClO₂ Boiling Pt: 92…93 °C Melting Pt: –81 °C Density: 1.136 Flash Pt: 10 °C(50 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00000644 CAS Number: 541-41-3 EINECS: 208-778-5 UN: 1182 ADR: "6.1",I Merck Index: 14,3784 |
Specification Test Results
Assay Minimum | 97 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...