5-Methyl-3-phenylisoxazole-4-carboxylic acid 99%
Supplier: Thermo Scientific Chemicals
L01142-14
L01142-22
CAAAL01142-14EA
72.21
CAD
CAAAL01142-14
CAAAL01142-22
5-Methyl-3-phenylisoxazole-4-carboxylic acid 99%
5-Methyl-3-phenylisoxazole-4-carboxylic acid
Purity: 99%
CAS No.: 1136-45-4
Molecular Formula: C11H9NO3
Formula Weight: 203.20
Odor: Odorless
Melting Point: 191-193°
Beilstein Reference: 164939
MDL No.: MFCD00003153
Formula:
C₁₁H₉NO₃ Melting Pt: 191…193 °C Storage Temperature: Ambient |
MDL Number:
MFCD00003153 CAS Number: 1136-45-4 EINECS: 214-497-9 |
Specification Test Results
Assay Minimum | 99 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...