Azido-PEG3-maleimide kit
Supplier: Thermo Scientific Chemicals
J65984-MB
J65984-MC
CAAAJ65984-MBEA
238.58
CAD
CAAAJ65984-MB
CAAAJ65984-MC
Azido-PEG3-maleimide kit
Azido-PEG3-maleimide
An azido containing reagent with PEG spacer and sulfhydryl reactivity. The compound is not stable upon prolonged storage and is sold as a kit.
Molecular Formula: C15H23N5O6
Formula Weight: 369.37
Storage Temperature: -30°C to -10°C
UN No.: 2735
MDL No.: MFCD22380764
Notes: Kit contains two components: a) 1.1 eq. of azido-PEG3-amine as an oil; b) 1.0 eq. of maleimide-NHS ester as a solid.
Formula:
C₁₅H₂₃N₅O₆ Storage Temperature: Refrigerator |
MDL Number:
MFCD22380764 UN: 2735 ADR: 8,III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...