Dimethyl-L-aspartate hydrochloride 98%
Supplier: Thermo Scientific Chemicals
Synonyms:
(S)-Aminosuccinic Acid Dimethyl Ester Hydrochloride, L-Asparaginic Acid Dimethyl Ester Hydrochloride, Dimethyl (S)-Aminosuccinate Hydrochloride, H-Asp(ome)-OMe HCl, Dimethyl L-Asparaginate Hydrochloride, H-Asp(OMe)-OMe.HCl, L-Aspartic Acid Dimethyl Ester Hydrochloride
Powder
Purity: 98%
CAS No.: 32213-95-9
Molecular Formula: C6H11NO4.HCl
Formula Weight: 197.62
Physical Form: Powder
Appearance: White to off-white
Melting Point: 115-120°
Optical Rotation: +15° (c=1 in methanol)
MDL No.: MFCD00038878
Formula:
C₆H₁₁NO₄ Melting Pt: 115…120 °C Storage Temperature: Ambient |
MDL Number:
MFCD00038878 CAS Number: 32213-95-9 EINECS: 250-957-4 |
Specification Test Results
Assay Minimum | 98 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...