N4-Benzoylcytidine 99%

Supplier: Thermo Scientific Chemicals

Synonyms: N-Benzoylcytidine

J65790-06 J65790-09 J65790-14
CAAAJ65790-06EA 124.98 CAD
CAAAJ65790-06 CAAAJ65790-09 CAAAJ65790-14
N4-Benzoylcytidine 99%
N4-Benzoylcytidine

Crystalline powder


Purity: 99% CAS No.: 13089-48-0 Molecular Formula: C16H17N3O6 Formula Weight: 347.32 Storage Temperature: 2°C to 8°C Physical Form: Crystalline powder Appearance: White to off-white Melting Point: 230-234° dec. Solubility: Soluble in 1N NaOH at 10mg/ml. Also soluble in methanol or DMSO Optical Rotation: +55° (c=0.1 in methanol) MDL No.: MFCD00010572

Formula: C₁₆H₁₇N₃O₆
Melting Pt: 230…234 °C (dec.)
Storage Temperature: Ambient
MDL Number: MFCD00010572
CAS Number: 13089-48-0

Order Now

Specification Test Results

Assay Minimum 99


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR