5-Bromo-4-chloro-3-indolyl-β-D-glucoside 99%

Supplier: Thermo Scientific Chemicals

Synonyms: 5-Bromo-4-chloro-3-indolyl beta-D-Glucopyranoside, X-Glc, 5-Bromo-4-chloro-3-(beta-D-glucopyranosyloxy)indole

J65677-03 J65677-ME
CAAAJ65677-03EA 183.28 CAD
CAAAJ65677-03 CAAAJ65677-06 CAAAJ65677-ME
5-Bromo-4-chloro-3-indolyl-β-D-glucoside 99%
5-Bromo-4-chloro-3-indolyl-beta-D-glucoside

Substrate for beta_x001A_-D-glucosidase


Crystalline powder


Applications: Substrate for beta-D-glucosidase

Formula: C₁₄H₁₅BrClNO₆
MW: 408.629 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD00063690
CAS Number: 15548-60-4
EINECS: 239-603-0

Order Now

Specification Test Results

Assay Minimum 99

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR