3-Azidopropylamine
Supplier: Thermo Scientific Chemicals
|
Warning
|
Synonyms:
3-Aminopropyl Azide, 3-Azido-1-propanamine, 3-Azido-1-propylamine, 1-Amino-3-azidopropane
J65668-03
J65668-MC
CAAAJ65668-03EA
982.76
CAD
CAAAJ65668-03
CAAAJ65668-MC
3-Azidopropylamine
3-Azidopropylamine
Azido-containing reagent with carbonyl and carboxyl reactivity
Liquid
Applications: Azido-containing reagent with carbonyl and carboxyl reactivity
Formula:
C₃H₈N₄ MW: 100.13 g/mol Flash Pt: 60 °C (140 °F) Storage Temperature: Refrigerator |
MDL Number:
MFCD11046568 CAS Number: 88192-19-2 UN: 1993 ADR: 3 III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...