Guanidinium hydrochloride for molecular biology

Supplier: Thermo Scientific Chemicals
Warning

J65661-22 J65661-36
CAAAJ65661-22EA 142.52 CAD
CAAAJ65661-22 CAAAJ65661-36
Guanidinium hydrochloride for molecular biology
Guanidine hydrochloride

A protein denaturant


Crystalline solid

Formula: NH₂C(=NH)NH₂·HCl
Melting Pt: 178…185 °C
Density: 1.34
Storage Temperature: Ambient
MDL Number: MFCD00013026
CAS Number: 50-01-1
EINECS: 200-002-3
Merck Index: 14,4562
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=da5a871cab977e6d9dc666b457732eb1e869a714&vid=b0d4fcb044b6106fce8e40448dbf2ce6e7077ad7&prid=web&clid=SAPDAM

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR