Azadibenzocyclooctyne-amine
Supplier: Thermo Scientific Chemicals
|
Warning
|
Synonyms:
DBCO-amine, 3-Amino-1-(11,12-didehydrodibenz[b,f]azocin-5(6H)-yl)-1-propanone, Dibenzocyclooctyne-amine
J65637-MB
J65637-MC
CAAAJ65637-MBEA
281.24
CAD
CAAAJ65637-MB
CAAAJ65637-MC
Azadibenzocyclooctyne-amine
Azadibenzocyclooctyne-amine
A carbonyl reactive reagent used to incorporate ADIBO into organic compounds, surfaces or particles
Caution: Notes: Heat sensitive. Incompatible with amines and azides.
Formula:
C₁₈H₁₆N₂O MW: 276.33 g/mol Storage Temperature: Refrigerator |
MDL Number:
MFCD22380759 CAS Number: 1255942-06-3 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...