Cephalothin sodium salt
Supplier: Thermo Scientific Chemicals
|
Danger
|
J65607-03
J65607-06
CAAAJ65607-03EA
165.9
CAD
CAAAJ65607-03
CAAAJ65607-06
Cephalothin sodium salt
Cephalothin sodium salt
A first-generation cephalosporin antibiotic
Powder
Applications: A first-generation cephalosporin antibiotic
Formula:
C₁₆H₁₅N₂NaO₆S₂ Storage Temperature: Ambient |
MDL Number:
MFCD00072025 CAS Number: 58-71-9 EINECS: 200-394-6 Merck Index: 14,1982 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...