Biebrich scarlet

Supplier: Thermo Scientific Chemicals

J65603-14 J65603-22
CAAAJ65603-14EA 48.02 CAD
CAAAJ65603-14 CAAAJ65603-22
Biebrich scarlet
Biebrich Scarlet

Powder


CAS: 4196-99-0 MDL: MFCD00003891

Formula: C₂₂H₁₄N₄Na₂O₇S₂
Melting Pt: 181…188 °C
Flash Pt: 300 °C(572 °F)
Storage Temperature: Ambient
MDL Number: MFCD00003891
CAS Number: 4196-99-0
EINECS: 224-084-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR