Water endotoxin-free
Supplier: Thermo Scientific Chemicals
J65589-AP
J65589-K2
CAAAJ65589-APEA
112.18
CAD
CAAAJ65589-AP
CAAAJ65589-K2
Water endotoxin-free
Water
Liquid
CAS: 7732-18-5
MDL: MFCD00011332
Formula:
H₂O Boiling Pt: 100 °C Melting Pt: 0 °C Density: 1.00 Storage Temperature: Ambient |
MDL Number:
MFCD00011332 CAS Number: 7732-18-5 EINECS: 231-791-2 Merck Index: 14,10039 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...