8-Bromoadenine
Supplier: Thermo Scientific Chemicals
|
Warning
|
J65582-03
J65582-MC
J65582-ME
CAAAJ65582-03EA
530.88
CAD
CAAAJ65582-03
CAAAJ65582-MC
CAAAJ65582-ME
8-Bromoadenine
8-Bromoadenine
Powder
CAS: 6974-78-3
MDL: MFCD00082518
Formula:
C₅H₄BrN₅ Melting Pt: >250 °C Storage Temperature: Ambient |
MDL Number:
MFCD00082518 CAS Number: 6974-78-3 EINECS: 230-225-1 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...