Ammonium 8-anilinonaphthalene-1-sulfonate 98%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: Ammonium 8-(phenylamino)naphthalene-1-sulphonate, Ammonium 8-(phenylamino)naphthalene-1-sulfonate, 8-Anilino-1-naphthalenesulphonic acid Ammonium salt, 8-Anilinonaphthalene-1-sulfonic acid ammonium salt, 8-Anilino-1-naphthalenesulfonic acid Ammonium salt, Ammonium 8-Anilino-1-naphthalenesulphonate, Ammonium 8-Anilino-1-naphthalenesulfonate, 8-Anilinonaphthalene-1-sulphonic acid ammonium salt, ANS-NH4

J65538-06 J65538-14 J65538-22
CAAAJ65538-06EA 79.95 CAD
CAAAJ65538-06 CAAAJ65538-14 CAAAJ65538-22
Ammonium 8-anilinonaphthalene-1-sulfonate 98%
Ammonium 8-anilinonaphthalene-1-sulphonate

Fluorescent probe for proteins on polyacrylamide gels.


Crystalline powder

Formula: C₁₆H₁₆N₂O₃S
MW: 316.37 g/mol
Melting Pt: 242…244 °C
Storage Temperature: Ambient
MDL Number: MFCD00012560
CAS Number: 28836-03-5
EINECS: 249-265-6

Order Now

Specification Test Results

Assay Minimum 98

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR