5-Aza-2'-deoxycytidine 98%
Supplier: Thermo Scientific Chemicals
|
Danger
|
Synonyms:
5-Aza-2′-deoxycytidine, Decitabine, 5-Aza-1-(2-deoxy-beta-D-ribofuranosyl)cytosine, 4-Amino-1-((2R,4S,5R)-4-hydroxy-5-(hydroxymethyl)tetrahydrofuran-2-yl)-1,3,5-triazin-2(1H)-one, 2′-Deoxy-5-azacytidine 4-Amino-1-(2-deoxy-β-D-ribofuranosyl)-1,3,5-triazin-2(1H)-one Decitabine, 4-Amino-1-(2-deoxy-beta-D-ribofuranosyl)-1,3,5-triazin-2(1H)-one
Powder
Purity: 98%
CAS No.: 2353-33-5
Molecular Formula: C8H12N4O4
Formula Weight: 228.21
Physical Form: Powder
Appearance: White to off-white
Beilstein Reference: 617982
MDL No.: MFCD00043011
Formula:
C₈H₁₂N₄O₄ MW: 228.21 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD00043011 CAS Number: 2353-33-5 EINECS: 219-089-4 Merck Index: 14,2853 |
Specification Test Results
Assay Minimum | 98 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...