5-Bromo-5-nitro-1,3-dioxane 98%, powder

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: Bromonitrodioxane, Bronidox L

J65443-06 J65443-14
CAAAJ65443-06EA 165.9 CAD
CAAAJ65443-06 CAAAJ65443-14
5-Bromo-5-nitro-1,3-dioxane 98%, powder
5-Bromo-5-nitro-1,3-dioxane

5-Bromo-5-nitro-1,3-dioxane is used as a stabilizer and preserving agent for biological molecules and solutions such as antibodies and antisera. Bronidox is used in a variety of rinse-off cosmetic. It can be used alone or in combination with methylisothiazolone.


Powder


Applications: Reactive against both Gram-positive and Gram-negative bacteria. Less toxic than sodium azide


Caution: Incompatible with strong oxidising agents.Storage must be tightly closed in well sealed container.

Formula: C₄H₆BrNO₄
MW: 212.00 g/mol
Melting Pt: 58…61 °C
Storage Temperature: Ambient
MDL Number: MFCD00101855
CAS Number: 30007-47-7
EINECS: 250-001-7

Order Now

Specification Test Results

Assay Minimum 98

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR