Artemisinin 98%

Supplier: Thermo Scientific Chemicals

J65406-03 J65406-06
CAAAJ65406-03EA 219.62 CAD
CAAAJ65406-03 CAAAJ65406-06
Artemisinin 98%
Artemisinin

Natural anti-malarial compound


Powder to crystals


Applications: Natural anti-malarial compound

Formula: C₁₅H₂₂O₅
Melting Pt: 156…157 °C
Storage Temperature: Ambient
MDL Number: MFCD00081057
CAS Number: 63968-64-9
Merck Index: 14,817

Order Now

Specification Test Results

Assay Minimum 98


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR