Dibenzocyclooctyne-maleimide
Supplier: Thermo Scientific Chemicals
|
Warning
|
Synonyms:
Azadibenzocyclooctyne-maleimide, ADIBO-maleimide, DBCO-maleimide
J65377-MB
J65377-MC
CAAAJ65377-MBEA
246.48
CAD
CAAAJ65377-MB
CAAAJ65377-MC
Dibenzocyclooctyne-maleimide
Dibenzocyclooctyne-maleimide
For incorporation of ADIBO functionality into sulfhydryl containing compounds
Formula:
C₂₅H₂₁N₃O₄ MW: 427.46 g/mol Storage Temperature: Refrigerator |
MDL Number:
MFCD22380750 CAS Number: 1395786-30-7 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...