Cafestol
Supplier: Thermo Scientific Chemicals
J65355-MC
J65355-MF
CAAAJ65355-MCEA
802.64
CAD
CAAAJ65355-MC
CAAAJ65355-MF
Cafestol
Cafestol
Natural extract from coffee beans which induces glutathione S-transferase
Powder
Applications: Natural extract from coffee beans which induces glutathione S-transferase
Formula:
C₂₀H₂₈O₃ MW: 316.441 g/mol Melting Pt: 156…158 °C Density: 1.23 Storage Temperature: Refrigerator |
MDL Number:
MFCD01075769 CAS Number: 469-83-0 Merck Index: 14,1634 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...