8-Anilino-1-naphthalenesulfonic acid 95%

Supplier: Thermo Scientific Chemicals
Warning

J65332-06 J65332-14 J65332-22
CAAAJ65332-06EA 72.68 CAD
CAAAJ65332-06 CAAAJ65332-14 CAAAJ65332-22
8-Anilino-1-naphthalenesulfonic acid 95%
8-Anilino-1-naphthalenesulphonic acid

Fluorescent probe for proteins on polyacrylamide gels


Powder

Formula: C₁₆H₁₃NO₃S
Melting Pt: 215…217 °C
Storage Temperature: Ambient
MDL Number: MFCD00003998
CAS Number: 82-76-8
EINECS: 201-438-7
Merck Index: 14,661

Order Now

Specification Test Results

Assay Minimum 95


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR