4-Amino-DL-phenylalanine
Supplier: Thermo Scientific Chemicals
Synonyms:
H-p-Amino-DL-Phe-OH hydrate
J65331-03
J65331-06
CAAAJ65331-03EA
139.67
CAD
CAAAJ65331-03
CAAAJ65331-06
4-Amino-DL-phenylalanine
4-Amino-DL-phenylalanine
Powder
CAS No.: 2922-41-0
Molecular Formula: C9H12N2O2
Formula Weight: 180.21
Storage Temperature: 2°C to 8°C
Physical Form: Powder
Appearance: White to brown
Melting Point: 228-230°
Solubility: Soluble in water
MDL No.: MFCD00007917
Formula:
C₉H₁₂N₂O₂ MW: 180.207 g/mol Melting Pt: 228…230 °C Storage Temperature: Ambient |
MDL Number:
MFCD00007917 CAS Number: 2922-41-0 EINECS: 220-869-1 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...