8-Amino-2-naphthalenesulfonic acid 95%
Supplier: Thermo Scientific Chemicals
|
Danger
|
J65304-14
J65304-22
CAAAJ65304-14EA
110.6
CAD
CAAAJ65304-14
CAAAJ65304-22
8-Amino-2-naphthalenesulfonic acid 95%
8-Amino-2-naphthalenesulphonic acid
Powder
CAS: 119-28-8
MDL: MFCD00004032
Formula:
C₁₀H₉NO₃S Melting Pt: >300 °C Storage Temperature: Ambient |
MDL Number:
MFCD00004032 CAS Number: 119-28-8 EINECS: 204-311-4 UN: 2585 ADR: "8",III Merck Index: 142,351 |
Specification Test Results
Assay Minimum | 95 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...