Cefazolin sodium salt
Supplier: Thermo Scientific Chemicals
|
Danger
|
J65274-03
J65274-06
CAAAJ65274-03EA
203.82
CAD
CAAAJ65274-03
CAAAJ65274-06
Cefazolin sodium salt
Cefazolin sodium salt
Inhibits bacterial cell wall synthesis
Powder
Applications: Inhibits bacterial cell wall synthesis
Formula:
C₁₄H₁₄N₈O₄S₃ Melting Pt: 170…172 °C Storage Temperature: Ambient |
MDL Number:
MFCD00056883 CAS Number: 27164-46-1 EINECS: 248-278-4 Merck Index: 14,1917 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...