Cefoperazone sodium salt
Supplier: Thermo Scientific Chemicals
|
Danger
|
J65185-03
J65185-06
J65185-14
CAAAJ65185-03EA
182.27
CAD
CAAAJ65185-03
CAAAJ65185-06
CAAAJ65185-14
Cefoperazone sodium salt
Cefoperazone sodium salt
For studying the expression, binding, and inhibition of penicillin-binding proteins
CAS No.: 62893-20-3
Molecular Formula: C25H26N9NaO8S2
Formula Weight: 667.65
Storage Temperature: 2°C to 8°C
Physical Form: Powder
Appearance: White to off-white
Solubility: Soluble in water
Beilstein Reference: 4902135
MDL No.: MFCD07793331
Applications: For studying the expression, binding, and inhibition of penicillin-binding proteins
Formula:
C₂₅H₂₆N₉NaO₈S₂ Storage Temperature: Ambient |
MDL Number:
MFCD07793331 CAS Number: 62893-20-3 EINECS: 263-751-5 Merck Index: 14,1930 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...