4-Amino-3-hydroxybutyric acid
Supplier: Thermo Scientific Chemicals
J65145-06
J65145-14
J65145-22
CAAAJ65145-06EA
97.33
CAD
CAAAJ65145-06
CAAAJ65145-14
CAAAJ65145-22
4-Amino-3-hydroxybutyric acid
4-Amino-3-hydroxybutyric acid
Reduces uptake of iodine by the thyroid glands in animals. Antispasmodic
Powder
Applications: Reduces uptake of iodine by the thyroid glands in animals. Antispasmodic
Formula:
C₄H₉NO₃ Melting Pt: 223 °C (dec.) Storage Temperature: Ambient |
MDL Number:
MFCD00008141 CAS Number: 924-49-2 EINECS: 213-106-9 Merck Index: 14,444 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...